Assay Details
| Gene Symbol: LOC100797326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254062.2 | NP_001240991.1 | 1 - 2 | 622 | 67 | |
| GenBank mRNA | BT093786.1 | - | 1 - 2 | 623 | 67 | |
More Information
Set Membership: |
|

LOC100797326
Genomic Map