Assay Details
| Gene Symbol: LOC100776352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254176.2 | NP_001241105.1 | 4 - 5 | 636 | 88 | |
| GenBank mRNA | BT095973.1 | - | 4 - 5 | 638 | 88 | |
More Information
Set Membership: |
|

LOC100776352
Genomic Map