Assay Details
| Gene Symbol: LOC100816293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254203.2 | NP_001241132.1 | 2 - 3 | 247 | 83 | |
| GenBank mRNA | BT098002.1 | - | 2 - 3 | 1193 | 83 | |
More Information
Set Membership: |
|

LOC100816293
Genomic Map