Assay Details
Gene Symbol: LOC100786093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254252.2 | NP_001241181.1 | 1 - 2 | 126 | 74 | |
GenBank mRNA | BT097228.1 | - | 1 - 2 | 128 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT097228.1, NM_001254252.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
