Assay Details
Gene Symbol: LOC100815605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254651.1 | NP_001241580.1 | 8 - 9 | 1005 | 97 | |
GenBank mRNA | BT097759.1 | - | 8 - 9 | 1005 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
