Assay Details
Gene Symbol: LOC100775215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255131.2 | NP_001242060.1 | 1 - 2 | 87 | 91 | |
GenBank mRNA | BT093906.1 | - | 1 - 2 | 89 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
