Assay Details
Gene Symbol: LOC100802375 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255236.2 | NP_001242165.1 | 2 - 3 | 501 | 106 | |
GenBank mRNA | BT097755.1 | - | 2 - 3 | 502 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
