Assay Details
Gene Symbol: LOC100790612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255291.1 | NP_001242220.1 | 3 - 4 | 386 | 66 | |
GenBank mRNA | BT089455.1 | - | 3 - 4 | 385 | 66 | |
BT095718.1 | - | 3 - 4 | 386 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
