Assay Details
| Gene Symbol: LOC100810842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255308.2 | NP_001242237.1 | 9 - 10 | 641 | 65 | |
| GenBank mRNA | BT096134.1 | - | 9 - 10 | 643 | 65 | |
More Information
Set Membership: |
|

LOC100810842
Genomic Map