Assay Details
Gene Symbol: LOC100801138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255405.1 | NP_001242334.1 | 5 - 6 | 636 | 104 | |
GenBank mRNA | BT097829.1 | - | 5 - 6 | 636 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
