Assay Details
Gene Symbol: LOC100801138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255405.1 | NP_001242334.1 | 8 - 9 | 978 | 96 | |
GenBank mRNA | BT097829.1 | - | 8 - 9 | 978 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
