Assay Details
| Gene Symbol: LOC100792444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255443.2 | NP_001242372.1 | 2 - 3 | 201 | 87 | |
| GenBank mRNA | BT097143.1 | - | 2 - 3 | 203 | 87 | |
More Information
Set Membership: |
|

LOC100792444
Genomic Map