Assay Details
| Gene Symbol: AP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255745.1 | NP_001242674.1 | 7 - 8 | 633 | 78 | |
| GenBank mRNA | EU287456.1 | - | 7 - 8 | 633 | 78 | |
| KF831241.1 | - | 7 - 8 | 633 | 78 | ||
More Information
Set Membership: |
|

AP1A
Genomic Map