Assay Details
Gene Symbol: LOC100808414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255839.1 | NP_001242768.1 | 3 - 4 | 389 | 106 | |
GenBank mRNA | BT099034.1 | - | 3 - 4 | 389 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
