Assay Details
Gene Symbol: PLDZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001288593.1 | NP_001275522.1 | 18 - 19 | 3063 | 80 | |
GenBank mRNA | KF542816.1 | - | 18 - 19 | 3063 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
