Assay Details
Gene Symbol: LOC100806689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289215.1 | NP_001276144.1 | 7 - 8 | 1157 | 85 | |
GenBank mRNA | AK287294.1 | - | 7 - 8 | 1229 | 85 | |
FJ594403.1 | - | 7 - 8 | 1157 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
