Assay Details
| Gene Symbol: LOC100792205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289303.1 | NP_001276232.1 | 1 - 2 | 263 | 62 | |
| GenBank mRNA | AK244417.1 | - | 1 - 2 | 264 | 62 | |
| AK286980.1 | - | 1 - 2 | 262 | 62 | ||
| BT097954.1 | - | 1 - 2 | 265 | 62 | ||
More Information
Set Membership: |
|

LOC100792205
Genomic Map