Assay Details
Gene Symbol: RCA03 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289312.1 | NP_001276241.1 | 1 - 2 | 46 | 73 | |
GenBank mRNA | AK246087.1 | - | 1 - 2 | 175 | 73 | |
GQ917181.1 | - | 1 - 2 | 46 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
