Assay Details
| Gene Symbol: RCA03 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289312.1 | NP_001276241.1 | 5 - 6 | 1304 | 83 | |
| GenBank mRNA | AK246087.1 | - | 5 - 6 | 1433 | 83 | |
| GQ917181.1 | - | 5 - 6 | 1304 | 83 | ||
More Information
Set Membership: |
|

RCA03
Genomic Map