Assay Details
| Gene Symbol: LOC100797902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK243884.1 | - | 4 - 5 | 379 | 71 | |
| AK286941.1 | - | 3 - 4 | 289 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100797902| Gene Symbol: LOC100797902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK243884.1 | - | 4 - 5 | 379 | 71 | |
| AK286941.1 | - | 3 - 4 | 289 | 71 | ||
Set Membership: |
|