Assay Details
| Gene Symbol: LOC100797902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK243884.1 | - | 9 - 10 | 904 | 115 | |
| AK286941.1 | - | 8 - 9 | 814 | 115 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100797902| Gene Symbol: LOC100797902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK243884.1 | - | 9 - 10 | 904 | 115 | |
| AK286941.1 | - | 8 - 9 | 814 | 115 | ||
Set Membership: |
|