Assay Details
Gene Symbol: LOC100811963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255711.1 | NP_001242640.1 | 1 - 2 | 431 | 62 | |
GenBank mRNA | BT098937.1 | - | 1 - 2 | 431 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
