Assay Details
| Gene Symbol: LOC100789743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255903.2 | NP_001242832.1 | 3 - 4 | 507 | 85 | |
| GenBank mRNA | BT097219.1 | - | 3 - 4 | 510 | 85 | |
More Information
Set Membership: |
|

LOC100789743
Genomic Map