Assay Details
Gene Symbol: LOC100805092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254560.2 | NP_001241489.1 | 2 - 3 | 337 | 104 | |
GenBank mRNA | BT096810.1 | - | 2 - 3 | 339 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
