Assay Details
| Gene Symbol: LOC100817791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255453.2 | NP_001242382.1 | 4 - 5 | 489 | 72 | |
| GenBank mRNA | BT098867.1 | - | 4 - 5 | 490 | 72 | |
| Gene Symbol: LOC100786514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK285325.1 | - | 4 - 5 | 72 | ||
More Information
Set Membership: |
|

LOC100817791
Genomic Map