Assay Details
| Gene Symbol: LOC100803909 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253077.1 | NP_001240006.1 | 4 | 624 | 167 | |
| GenBank mRNA | AB550124.1 | - | 4 | 624 | 167 | |
More Information
Set Membership: |
|

LOC100803909
Genomic Map