Assay Details
Gene Symbol: LOC100804444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253197.2 | NP_001240126.1 | 6 | 908 | 89 | |
GenBank mRNA | AK286785.1 | - | 6 | 825 | 89 | |
BT094926.1 | - | 1 | 168 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
