Assay Details
| Gene Symbol: LOC100780934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254021.2 | NP_001240950.1 | 1 | 117 | 91 | |
| GenBank mRNA | BT095576.1 | - | 1 | 120 | 91 | |
More Information
Set Membership: |
|

LOC100780934
Genomic Map