Assay Details
| Gene Symbol: LOC100791447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254121.2 | NP_001241050.1 | 2 | 522 | 61 | |
| GenBank mRNA | AK286464.1 | - | 2 | 518 | 61 | |
| BT096175.1 | - | 2 | 525 | 61 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				

LOC100791447
Genomic Map