Assay Details
Gene Symbol: LOC100779203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254384.2 | NP_001241313.1 | 8 | 1018 | 137 | |
GenBank mRNA | AK286651.1 | - | 8 | 1019 | 137 | |
BT096730.1 | - | 8 | 1020 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
