Assay Details
| Gene Symbol: LOC100800967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254400.2 | NP_001241329.1 | 3 | 515 | 133 | |
| GenBank mRNA | BT096531.1 | - | 3 | 517 | 133 | |
More Information
Set Membership: |
|

LOC100800967
Genomic Map