Assay Details
| Gene Symbol: LOC100775676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254685.2 | NP_001241614.1 | 3 | 1216 | 109 | |
| GenBank mRNA | BT097096.1 | - | 3 | 1218 | 109 | |
More Information
Set Membership: |
|

LOC100775676
Genomic Map