Assay Details
| Gene Symbol: LOC100776238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255125.2 | NP_001242054.1 | 1 | 125 | 101 | |
| GenBank mRNA | BT094360.1 | - | 1 | 127 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT094360.1, NM_001255125.2 transcripts.
Set Membership: |
|

LOC100776238
Genomic Map