Assay Details
Gene Symbol: LOC100805710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255270.2 | NP_001242199.1 | 1 | 121 | 120 | |
GenBank mRNA | BT097515.1 | - | 1 | 124 | 120 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT097515.1, NM_001255270.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
