Assay Details
Gene Symbol: LOC100794961 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255282.2 | NP_001242211.1 | 1 | 119 | 82 | |
GenBank mRNA | BT093964.1 | - | 1 | 121 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT093964.1, NM_001255282.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
