Assay Details
Gene Symbol: LOC100775804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255440.2 | NP_001242369.1 | 1 | 221 | 88 | |
GenBank mRNA | BT098847.1 | - | 1 | 223 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT098847.1, NM_001255440.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
