Assay Details
| Gene Symbol: LOC100804015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255655.2 | NP_001242584.1 | 1 | 121 | 99 | |
| GenBank mRNA | BT096410.1 | - | 1 | 123 | 99 | |
More Information
Set Membership: |
|

LOC100804015
Genomic Map