Assay Details
| Gene Symbol: LOC100785878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254485.2 | NP_001241414.1 | 4 | 913 | 104 | |
| GenBank mRNA | AK245791.1 | - | 4 | 914 | 104 | |
| BT096645.1 | - | 4 | 915 | 104 | ||
More Information
Set Membership: |
|

LOC100785878
Genomic Map