Assay Details
Gene Symbol: LOC100810230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254658.1 | NP_001241587.1 | 3 | 525 | 87 | |
GenBank mRNA | BT094034.1 | - | 3 | 525 | 87 |
Gene Symbol: LOC100778110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK244654.1 | - | 3 | 531 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
