Assay Details
| Gene Symbol: LOC100781853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255063.1 | NP_001241992.1 | 5 | 617 | 110 | |
| GenBank mRNA | AK287321.1 | - | 5 | 606 | 110 | |
| BT093242.1 | - | 5 | 617 | 110 | ||
More Information
Set Membership: |
|

LOC100781853
Genomic Map