Assay Details
| Gene Symbol: LOC100788368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255538.2 | NP_001242467.1 | 5 | 1122 | 68 | |
| GenBank mRNA | BT097631.1 | - | 5 | 1124 | 68 | |
More Information
Set Membership: |
|

LOC100788368
Genomic Map