Assay Details
| Gene Symbol: LOC100803810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255853.1 | NP_001242782.1 | 1 | 215 | 103 | |
| GenBank mRNA | BT095978.1 | - | 1 | 215 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT095978.1, NM_001255853.1 transcripts.
Set Membership: |
|

LOC100803810
Genomic Map