Assay Details
Gene Symbol: Cadm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004856630.3 | XP_004856687.1 | 11 - 12 | 1334 | 79 | |
XM_004856631.3 | XP_004856688.1 | 10 - 11 | 1278 | 79 | ||
XM_004856632.3 | XP_004856689.1 | 10 - 11 | 1257 | 79 | ||
XM_004856633.3 | XP_004856690.1 | 9 - 10 | 1244 | 79 | ||
XM_004856634.3 | XP_004856691.1 | 9 - 10 | 1201 | 79 | ||
XM_004856635.3 | XP_004856692.1 | 8 - 9 | 1168 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
