Assay Details
Gene Symbol: Mtcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004875182.3 | XP_004875239.1 | 2 - 3 | 3205 | 75 | |
XM_004875183.3 | XP_004875240.1 | 3 - 4 | 408 | 75 | ||
XM_013069449.2 | XP_012924903.1 | 3 - 4 | 513 | 75 | ||
XM_021252450.1 | XP_021108109.1 | 4 - 5 | 648 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
