Assay Details
Gene Symbol: Yipf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839271.2 | XP_004839328.1 | 5 - 6 | 882 | 69 | |
XM_004839272.2 | XP_004839329.1 | 5 - 6 | 860 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
