Assay Details
| Gene Symbol: Ube2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247135.1 | XP_021102794.1 | 1 | 727 | 123 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2e3| Gene Symbol: Ube2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247135.1 | XP_021102794.1 | 1 | 727 | 123 | |
Set Membership: |
|