Assay Details
| Gene Symbol: Pea15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858725.3 | XP_004858782.1 | 3 - 4 | 485 | 73 | |
| XM_013077481.2 | XP_012932935.1 | 3 - 4 | 392 | 73 | ||
More Information
Set Membership: |
|

Pea15
Genomic Map