Assay Details
| Gene Symbol: Kcnc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866833.2 | XP_004866890.1 | 1 - 2 | 1636 | 82 | |
| XM_013066345.2 | XP_012921799.1 | 1 - 2 | 1636 | 82 | ||
| XM_013066346.2 | XP_012921800.1 | 1 - 2 | 1636 | 82 | ||
| XM_021246031.1 | XP_021101690.1 | 1 - 2 | 1636 | 82 | ||
| XM_021246032.1 | XP_021101691.1 | 1 - 2 | 1636 | 82 | ||
More Information
Set Membership: |
|

Kcnc3
Genomic Map