Assay Details
| Gene Symbol: Dcun1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874023.2 | XP_004874080.1 | 2 - 3 | 490 | 61 | |
| XM_021251568.1 | XP_021107227.1 | 1 - 2 | 93 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021251568.1 transcripts.
Set Membership: |
|

Dcun1d5
Genomic Map