Assay Details
| Gene Symbol: Chtop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871426.3 | XP_004871483.1 | 2 - 3 | 476 | 67 | |
| XM_004871427.3 | XP_004871484.1 | 2 - 3 | 103 | 67 | ||
| XM_004871429.3 | XP_004871486.1 | 2 - 3 | 440 | 67 | ||
| XM_004871430.3 | XP_004871487.1 | 2 - 3 | 483 | 67 | ||
| XM_013067981.2 | XP_012923435.1 | 2 - 3 | 462 | 67 | ||
| XM_013067982.2 | XP_012923436.1 | 2 - 3 | 483 | 67 | ||
| XM_021249516.1 | XP_021105175.1 | 2 - 3 | 278 | 67 | ||
| XM_021249517.1 | XP_021105176.1 | 2 - 3 | 477 | 67 | ||
| XM_021249518.1 | XP_021105177.1 | 2 - 3 | 433 | 67 | ||
| XM_021249520.1 | XP_021105179.1 | 2 - 3 | 483 | 67 | ||
More Information
Set Membership: |
|

Chtop
Genomic Map